Learn More
Medchemexpress LLC Fluorogenic dibenzocyclooctyne | 1407523-31-2 | MFCD32645582 | 99.3% | 288.30 | C19H12O3 | 10 MG
SDP
Catalog No. 5000297185
Encompass_Preferred
Fl-DIBO is a fluorogenic dibenzocyclooctyne reagent used for bioorthogonal labeling of azide-containing molecules. It reacts rapidly with azides to form highly fluorescent products (excitation 363 nm, emission 469 nm), enabling sensitive labeling with low background.
  • Fluorogenic probe for azide compounds enabling sensitive detection
  • Rapid click chemistry reaction producing strong fluorescence (excitation 363 nm; emission 469 nm)
  • High purity (~99.3%) and defined molecular weight (288.30)
  • Available in small research quantities such as 10 mg
  • Store at 4°C, protect from light; in solvent store at -80°C for long-term stability
Catalog No. 50-002-97185 Supplier Medchemexpress LLC Supplier No. HYD150610MG
Fl-DIBO is a fluorogenic dibenzocyclooctyne reagent used for bioorthogonal labeling of azide-containing molecules. It reacts rapidly with azides to form highly fluorescent products (excitation 363 nm, emission 469 nm), enabling sensitive labeling with low background.
  • Fluorogenic probe for azide compounds enabling sensitive detection
  • Rapid click chemistry reaction producing strong fluorescence (excitation 363 nm; emission 469 nm)
  • High purity (~99.3%) and defined molecular weight (288.30)
  • Available in small research quantities such as 10 mg
  • Store at 4°C, protect from light; in solvent store at -80°C for long-term stability