Learn More
Bachem Mca-(Asn670,Leu671)-Amyloid Beta/A4 Protein Precursor770 (667-675)-Lys(Dnp) Ammonium acetate
SDP
Supplier:  Bachem 4029476.0001
Encompass_Preferred
Mca-(Asn670,Leu671)-Amyloid Beta/A4 Protein Precursor770 (667-675)-Lys(Dnp) Ammonium acetate, Mca-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Lys(Dnp)-OH, 1802078-31-4, C68H88N14O27, Mr 1533.53, 1mg. Mca-(Asn670,Leu671)-APP770 (667-675)-Lys(Dnp). Fluorogenic (FRET) substrate for pro-memapsin-2 containing the b-secretase site of the Swedish mutation of APP, SEVNLDAEF.
Catalog No. 50-194-6612