Learn More
Bachem H-Arg-Glu(EDANS)-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Lys(DABCYL)-Arg-OH
SDP
Supplier:  Bachem 4033536.0001
Encompass_Preferred
Arg-Glu(EDANS)-(Asn670,Leu671)-Amyloid b/A4 Protein Precursor770 (668-675)-Lys(DABCYL)-Arg Trifluoroacetate, H-Arg-Glu(EDANS)-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Lys(DABCYL)-Arg-OH, 310427-94-2, C91H129N25O25S, Mr 2005.25, 1mg. Arg-Glu(EDANS)-(Asn670,Leu671)-APP770 (668-675)-Lys(DABCYL)-Arg. Fluorogenic (FRET) substrate for pro-memapsin-2 containing the b-secretase site EVNLDAEF of the Swedish mutation of APP. The kinetic parameters at pH 4.5 are Km = 5.4 uM and kcat = 0.24 min-1.
Catalog No. 50-194-6625