Learn More
Bachem H-Ala-AFC
SDP
Supplier:  Bachem 4009776.1000
Encompass_Preferred
H-Ala-AFC Trifluoroacetate, 126910-31-4, C13H11F3N2O3, Mr 300.24, 1g. Fluorogenic substrate for microsomal alanine aminopeptidase, excitation at 400 nm, emission at 505 nm.
Catalog No. 50-194-7031