Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorogestone acetate | 2529-45-5 | 99.6% | 406.49 g·mol⁻¹ | C23H31FO5 | 50MG
SDP

Catalog No. 5000239364
Encompass_Preferred

Fluorogestone acetate is a synthetic progestin used in veterinary and research settings to inhibit ovulation and synchronize estrous cycles. Supplied as a solid, high-purity reagent, it is characterized by formula C23H31FO5 and a molecular weight of 406.49 g·mol⁻¹; handle and store sealed away from moisture and light as recommended in the SDS.

  • High purity: 99.6%.
  • Molecular formula: C23H31FO5.
  • Molecular weight: 406.49 g·mol⁻¹.
  • Available in multiple pack sizes including 50 mg.
  • Recommended storage: 4°C sealed; in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-39364 Supplier Medchemexpress LLC Supplier No. HY12561150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogestone acetate is a synthetic progestin used in veterinary and research settings to inhibit ovulation and synchronize estrous cycles. Supplied as a solid, high-purity reagent, it is characterized by formula C23H31FO5 and a molecular weight of 406.49 g·mol⁻¹; handle and store sealed away from moisture and light as recommended in the SDS.

  • High purity: 99.6%.
  • Molecular formula: C23H31FO5.
  • Molecular weight: 406.49 g·mol⁻¹.
  • Available in multiple pack sizes including 50 mg.
  • Recommended storage: 4°C sealed; in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.