Learn More
Medchemexpress LLC Fluorogestone acetate | 2529-45-5 | MFCD00243094 | 99.6% | 406.49 | C23H31FO5 | 100MG
SDP
Catalog No. 5000239387
Encompass_Preferred
Fluorogestone acetate is a synthetic progesterone analog used to inhibit ovulation and synchronize the estrous cycle in cattle by suppressing luteinizing hormone (LH) release. It is supplied as a high-purity reagent in weighed milligram quantities for research and veterinary reproductive management applications.
  • Inhibits ovulation and synchronizes the estrous cycle in cattle.
  • Acts by suppressing luteinizing hormone (LH) release.
  • High purity: 99.6%.
  • Molecular formula: C23H31FO5; molecular weight: 406.49.
  • Available in multiple package sizes, including 100 mg.
  • Provided in weighed quantities suitable for formulation and analytical use.
Catalog No. 50-002-39387 Supplier Medchemexpress LLC Supplier No. HY125611100MG
Fluorogestone acetate is a synthetic progesterone analog used to inhibit ovulation and synchronize the estrous cycle in cattle by suppressing luteinizing hormone (LH) release. It is supplied as a high-purity reagent in weighed milligram quantities for research and veterinary reproductive management applications.
  • Inhibits ovulation and synchronizes the estrous cycle in cattle.
  • Acts by suppressing luteinizing hormone (LH) release.
  • High purity: 99.6%.
  • Molecular formula: C23H31FO5; molecular weight: 406.49.
  • Available in multiple package sizes, including 100 mg.
  • Provided in weighed quantities suitable for formulation and analytical use.