Learn More
Medchemexpress LLC Fluorogestone acetate | 2529-45-5 | MFCD00243094 | 99.6% | 406.49 | C23H31FO5 | 5MG

Supplier: Medchemexpress LLC HY1256115MG
Fluorogestone acetate is a synthetic progesterone analogue used primarily in veterinary reproductive management to inhibit ovulation and synchronize the estrous cycle by suppressing luteinizing hormone release. It is supplied as a high-purity chemical for research and formulation applications.
- High purity (99.6%).
- Molecular formula C23H31FO5, molecular weight 406.49.
- CAS number 2529-45-5.
- Available sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
- Used to inhibit ovulation and synchronize estrous cycles in cattle.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.