Learn More
Medchemexpress LLC Fluorogestone acetate | 2529-45-5 | MFCD00243094 | 99.6% | 406.49 g/mol | C23H31FO5 | 10MG
SDP
Catalog No. 5000239625
Encompass_Preferred
Fluorogestone acetate is a synthetic progesterone analogue used in research and veterinary applications to inhibit ovulation and synchronize estrous cycles. It is supplied as a solid, high-purity compound suitable for laboratory assays, in vivo studies, and formulation work. Handle with standard laboratory precautions and protect from moisture and light during storage.
Catalog No. 50-002-39625 Supplier Medchemexpress LLC Supplier No. HY12561110MG
Add to Cart
Add to Cart
Fluorogestone acetate is a synthetic progesterone analogue used in research and veterinary applications to inhibit ovulation and synchronize estrous cycles. It is supplied as a solid, high-purity compound suitable for laboratory assays, in vivo studies, and formulation work. Handle with standard laboratory precautions and protect from moisture and light during storage.