Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorogestone acetate | 2529-45-5 | MFCD00243094 | 99.6% | 406.49 g/mol | C23H31FO5 | 25MG
SDP

Catalog No. 5000237862
Encompass_Preferred

Fluorogestone acetate is a synthetic progesterone derivative used to inhibit ovulation and synchronize the estrous cycle in cattle. It is intended for research and veterinary applications and is characterized by defined purity and chemical properties.

  • Purity: 99.6%.
  • Molecular formula: C23H31FO5.
  • Molecular weight: 406.49 g/mol.
  • Application: inhibits ovulation and synchronizes estrous cycles in cattle.
  • Packaging: available in multiple small pack sizes for dosing flexibility.
  • Intended use: for research use and veterinary studies, not for human therapeutic use.

Catalog No. 50-002-37862 Supplier Medchemexpress LLC Supplier No. HY12561125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogestone acetate is a synthetic progesterone derivative used to inhibit ovulation and synchronize the estrous cycle in cattle. It is intended for research and veterinary applications and is characterized by defined purity and chemical properties.

  • Purity: 99.6%.
  • Molecular formula: C23H31FO5.
  • Molecular weight: 406.49 g/mol.
  • Application: inhibits ovulation and synchronizes estrous cycles in cattle.
  • Packaging: available in multiple small pack sizes for dosing flexibility.
  • Intended use: for research use and veterinary studies, not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.