Learn More
Medchemexpress LLC Fluorol Yellow 088 | 81-37-8 | ≥98.0% | 250 MG
SDP
Catalog No. 5000414241
Encompass_Preferred
Fluorol Yellow 088 is a fluorescent dye utilized as a biological material or an organic compound for life science research. It is suitable for lipid staining in plant tissues, particularly for the fluorescence imaging of suberized cell walls and hydrophobic structures. The dye has an excitation wavelength of 365 nm and an emission wavelength of ≥420 nm.
  • Strong staining stability, resisting precipitation in aqueous media.
  • High lipid specificity, leading to high-contrast imaging.
  • Suitable for large-scale scanning (rapid screening under low magnification).
  • Primarily used for lipid staining in plant tissues, especially for the visualization of suberin lamellae, endodermis, and exodermis.
Catalog No. 50-004-14241 Supplier Medchemexpress LLC Supplier No. 250MGHYD0286
Fluorol Yellow 088 is a fluorescent dye utilized as a biological material or an organic compound for life science research. It is suitable for lipid staining in plant tissues, particularly for the fluorescence imaging of suberized cell walls and hydrophobic structures. The dye has an excitation wavelength of 365 nm and an emission wavelength of ≥420 nm.
  • Strong staining stability, resisting precipitation in aqueous media.
  • High lipid specificity, leading to high-contrast imaging.
  • Suitable for large-scale scanning (rapid screening under low magnification).
  • Primarily used for lipid staining in plant tissues, especially for the visualization of suberin lamellae, endodermis, and exodermis.