Learn More
Santa Cruz Biotechnology Fluorol Yellow 088, 81-37-8, MFCD00083335, 5 g
Supplier:  Santa Cruz Biotechnology SC215052
Encompass
Molecular Weight: 296.36, Molecular Formula: C22H16O, ≥95%, Store at room temperature, MP: 190.26° C, BP: ~512.5° C, Synonym: 2,8-Dimethylnaphtho[3,2,1-kl]xanthene; Solvent Green 4.
Catalog No. NC0781160