Learn More
Sigma Aldrich Fine Chemicals Biosciences fluorolubeR1EA
Catalog No. 502905043
Encompass
for IR sample mulls. 1 oz. tube.
Catalog No. 50-290-5043 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1235521EA
for IR sample mulls. 1 oz. tube.