Learn More
Starna Cells Inc FLUOROMETER CUVETTE
SDP
Catalog No. NC9892739
Encompass_Preferred
Fluorometer Cuvette, Path Length: 10mm, Exterior Dimension: 12.5L x 12.5W x 25mmH, Interior Dimension: 10L x 2mmW, Nominal Volume: 0.400 mL, Window Material: Spectrosil UV Quartz windows, Useable Range: 170 to 2700nm, Polished Windows: 4
Catalog No. NC9892739 Supplier Starna Cells Inc Supplier No. 17FQ10
Fluorometer Cuvette, Path Length: 10mm, Exterior Dimension: 12.5L x 12.5W x 25mmH, Interior Dimension: 10L x 2mmW, Nominal Volume: 0.400 mL, Window Material: Spectrosil UV Quartz windows, Useable Range: 170 to 2700nm, Polished Windows: 4