Learn More
Starna Cells Inc FLUOROMETER MICRO SQUARE CELL
SDP
Catalog No. NC9876151
Encompass_Preferred
Square Micro Fluorometer Cell, open top,Window material:Spectrosil Quartz or equivalent, useable range:170 to 2700 nm, Path Length:3 mm, Exterior (W x L x H): 5.5 x 5.5 x 30 mm, Interior (W x L): 3 x 3 mm, Nominal Volume : 0.315ml, Number P
Catalog No. NC9876151 Supplier Starna Cells Inc Supplier No. 33.45Q3
Square Micro Fluorometer Cell, open top,Window material:Spectrosil Quartz or equivalent, useable range:170 to 2700 nm, Path Length:3 mm, Exterior (W x L x H): 5.5 x 5.5 x 30 mm, Interior (W x L): 3 x 3 mm, Nominal Volume : 0.315ml, Number P