Learn More
Cayman Chemical Fluorometholon 100mg
SDP
Catalog No. 502896831
Encompass_Preferred
A synthetic glucocorticoid that binds to glucocorticoid receptors (IC50 = 1.5 nM; Kd = 2.8 nM); decreases survival of cultured corneal epithelial cells and inhibits their migration in a scratch assay following 12 hours of treatment (0.01%)
Catalog No. 50-289-6831 Supplier Cayman Chemical Supplier No. 9000157100 MG
A synthetic glucocorticoid that binds to glucocorticoid receptors (IC50 = 1.5 nM; Kd = 2.8 nM); decreases survival of cultured corneal epithelial cells and inhibits their migration in a scratch assay following 12 hours of treatment (0.01%)