Learn More
Cayman Chemical Fluorometholone, Purity: ≥98%, CAS: 426-13-1, 50 mg
SDP
Catalog No. 502896561
Encompass_Preferred
A synthetic glucocorticoid that binds to glucocorticoid receptors (IC50 = 1.5 nM; Kd = 2.8 nM); decreases survival of cultured corneal epithelial cells and inhibits their migration in a scratch assay following 12 hours of treatment (0.01%)
Catalog No. 50-289-6561 Supplier Cayman Chemical Supplier No. 900015750 MG
A synthetic glucocorticoid that binds to glucocorticoid receptors (IC50 = 1.5 nM; Kd = 2.8 nM); decreases survival of cultured corneal epithelial cells and inhibits their migration in a scratch assay following 12 hours of treatment (0.01%)