Learn More
Medchemexpress LLC HY-B1893 25mg Medchemexpress, Fluorometholone CAS:426-13-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB189325MG
Encompass_Preferred
Medchemexpress, HY-B1893 25mg Fluorometholone CAS:426-13-1 Fluorometholone, a synthetic glucocorticoid, is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic properties. Fluorometholone can be used for the research of dry eye. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1241