Learn More
Medchemexpress LLC Fluorometholone | 426-13-1 | 99.4% | 1 ML
SDP
Catalog No. 5000411397
Encompass_Preferred
Fluorometholone, a synthetic glucocorticoid, is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic properties. It can be used for the research of dry eye.
  • Synthetic glucocorticoid
  • Glucocorticoid receptor agonist
  • Anti-inflammatory properties
  • Anti-allergic properties
  • Useful for dry eye research
Catalog No. 50-004-11397 Supplier Medchemexpress LLC Supplier No. HYB189310MM/1ML
Add to Cart
Add to Cart
Fluorometholone, a synthetic glucocorticoid, is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic properties. It can be used for the research of dry eye.
  • Synthetic glucocorticoid
  • Glucocorticoid receptor agonist
  • Anti-inflammatory properties
  • Anti-allergic properties
  • Useful for dry eye research