Learn More
Medchemexpress LLC Fluorometholone | 426-13-1 | 99.4% | 50 MG
SDP
Catalog No. 5000409645
Encompass_Preferred
Fluorometholone is a synthetic glucocorticoid and a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic properties. It can be used for the research of dry eye. This product is for research use only and not sold to patients.
  • Synthetic glucocorticoid
  • Glucocorticoid receptor agonist
  • Possesses anti-inflammatory and anti-allergic properties
  • Can be used for the research of dry eye
  • Increases the gene expression of MUC1, MUC4, MUC16 and MUC19 in conjunctival and corneal epithelial cells through activation of glucocorticoid receptors
Catalog No. 50-004-09645 Supplier Medchemexpress LLC Supplier No. HYB189350MG
Fluorometholone is a synthetic glucocorticoid and a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic properties. It can be used for the research of dry eye. This product is for research use only and not sold to patients.
  • Synthetic glucocorticoid
  • Glucocorticoid receptor agonist
  • Possesses anti-inflammatory and anti-allergic properties
  • Can be used for the research of dry eye
  • Increases the gene expression of MUC1, MUC4, MUC16 and MUC19 in conjunctival and corneal epithelial cells through activation of glucocorticoid receptors