Learn More
Apexbio Technology LLC Fluorometholone Acetate 3801-06-7 50mg
SDP
Catalog No. 5000094130
Encompass_Preferred
Fluorometholone Acetate (CAS 3801-06-7) is a synthetic glucocorticoid corticosteroid designed for the modulation of inflammatory responses Acting through binding to the glucocorticoid receptor it regulates the transcription of anti-inflammatory proteins while suppressing pro-inflammatory gene expression This compound is extensively utilized in ophthalmic research for modeling and investigating steroid-responsive inflammatory processes in ocular tissues Its application facilitates the study of corticosteroid-mediated pathways in eye inflammation and aids in the evaluation of anti-inflammatory interventions in preclinical settings
Catalog No. 50-000-94130 Supplier Apexbio Technology LLC Supplier No. B176250
Fluorometholone Acetate (CAS 3801-06-7) is a synthetic glucocorticoid corticosteroid designed for the modulation of inflammatory responses Acting through binding to the glucocorticoid receptor it regulates the transcription of anti-inflammatory proteins while suppressing pro-inflammatory gene expression This compound is extensively utilized in ophthalmic research for modeling and investigating steroid-responsive inflammatory processes in ocular tissues Its application facilitates the study of corticosteroid-mediated pathways in eye inflammation and aids in the evaluation of anti-inflammatory interventions in preclinical settings