Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorometholone acetate | 3801-06-7 | 98.5% | 1 ML
SDP

Supplier:  Medchemexpress LLC HYB147110MM/1ML

Encompass_Preferred

Fluorometholone acetate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. It potently inhibits carbonic anhydrase (CA) with IC50s of 2.18 μM and 17.5 μM for hCA-I and hCA-II, respectively. It has anti-inflammatory effects and shows potential for external ocular inflammation research.

  • Molecular weight of 418.50
  • Chemical formula C24H31FO5
  • Appears as a solid
  • White to off-white color
  • Soluble in DMSO at 100 mg/mL (238.95 mM)
  • Store powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Potent inhibitor of carbonic anhydrase (CA)
  • Exhibits anti-inflammatory effects
  • Potential for external ocular inflammation research
  • Ki values for hCA-I and hCA-II are 1.044 μM and 9.98 μM, respectively
  • Reduced polymorphonuclear leukocytes in the cornea by an average of 47% in vivo studies

Catalog No. 50-004-11161


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.