Learn More
Medchemexpress LLC Fluorometholone acetate | 3801-06-7 | 98.5% | 100 MG
SDP
Catalog No. 5000412223
Encompass_Preferred
Fluorometholone acetate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. It is a potent inhibitor of carbonic anhydrase (CA) with IC50 values of 2.18 μM for hCA-I and 17.5 μM for hCA-II. It also has an anti-inflammatory effect and has potential for research into external ocular inflammation.
  • Synthetic glucocorticoid corticosteroid
  • Corticosteroid ester
  • Potent inhibitor of carbonic anhydrase (CA)
  • Anti-inflammatory effect
  • Potential for external ocular inflammation research
  • For research use only
Catalog No. 50-004-12223 Supplier Medchemexpress LLC Supplier No. HYB1471100MG
Fluorometholone acetate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. It is a potent inhibitor of carbonic anhydrase (CA) with IC50 values of 2.18 μM for hCA-I and 17.5 μM for hCA-II. It also has an anti-inflammatory effect and has potential for research into external ocular inflammation.
  • Synthetic glucocorticoid corticosteroid
  • Corticosteroid ester
  • Potent inhibitor of carbonic anhydrase (CA)
  • Anti-inflammatory effect
  • Potential for external ocular inflammation research
  • For research use only