Learn More
Medchemexpress LLC Fluorometholone acetate | 3801-06-7 | 98.5% | 25 MG
SDP
Catalog No. 5000413890
Encompass_Preferred
Fluorometholone acetate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. It potently inhibits carbonic anhydrase (CA) and has anti-inflammatory effects, making it suitable for external ocular inflammation research.
  • Potently inhibits carbonic anhydrase (CA)
  • Inhibits hCA-I with IC50 of 2.18 μM
  • Inhibits hCA-II with IC50 of 17.5 μM
  • Used in inflammation/immunology research
  • Appears as a white to off-white solid
Catalog No. 50-004-13890 Supplier Medchemexpress LLC Supplier No. 25MGHYB1471
Fluorometholone acetate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. It potently inhibits carbonic anhydrase (CA) and has anti-inflammatory effects, making it suitable for external ocular inflammation research.
  • Potently inhibits carbonic anhydrase (CA)
  • Inhibits hCA-I with IC50 of 2.18 μM
  • Inhibits hCA-II with IC50 of 17.5 μM
  • Used in inflammation/immunology research
  • Appears as a white to off-white solid