Learn More
Medchemexpress LLC Fluorometholone acetate | 3801-06-7 | 98.5% | 50MG
SDP
Catalog No. 5000411895
Encompass_Preferred
Fluorometholone acetate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. It potently inhibits carbonic anhydrase (CA) with IC50s of 2.18 μM and 17.5 μM for hCA-I and hCA-II, respectively. Fluorometholone acetate has an anti-inflammatory effect and has the potential for external ocular inflammation research.
  • Synthetic glucocorticoid corticosteroid
  • Potent carbonic anhydrase inhibitor
  • Exhibits anti-inflammatory effects
  • Potential for external ocular inflammation research
Catalog No. 50-004-11895 Supplier Medchemexpress LLC Supplier No. HYB147150MG
Fluorometholone acetate is a synthetic glucocorticoid corticosteroid and a corticosteroid ester. It potently inhibits carbonic anhydrase (CA) with IC50s of 2.18 μM and 17.5 μM for hCA-I and hCA-II, respectively. Fluorometholone acetate has an anti-inflammatory effect and has the potential for external ocular inflammation research.
  • Synthetic glucocorticoid corticosteroid
  • Potent carbonic anhydrase inhibitor
  • Exhibits anti-inflammatory effects
  • Potential for external ocular inflammation research