Learn More
Medchemexpress LLC Fluorometholone (Standard) | 426-13-1 | MFCD00056461 | 98.8% | 5 MG
SDP
Catalog No. 5000412264
Encompass_Preferred
Fluorometholone (Standard) is the analytical standard of Fluorometholone, a synthetic glucocorticoid. It acts as a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic properties. This product is intended for research and analytical applications, including research on dry eye.
  • Analytical standard
  • Glucocorticoid receptor agonist
  • Anti-inflammatory properties
  • Anti-allergic properties
  • Used in qualitative, quantitative and methodological research experiments
  • Applications include HPLC, GC, and MS
Catalog No. 50-004-12264 Supplier Medchemexpress LLC Supplier No. HYB1893R5MG
Fluorometholone (Standard) is the analytical standard of Fluorometholone, a synthetic glucocorticoid. It acts as a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic properties. This product is intended for research and analytical applications, including research on dry eye.
  • Analytical standard
  • Glucocorticoid receptor agonist
  • Anti-inflammatory properties
  • Anti-allergic properties
  • Used in qualitative, quantitative and methodological research experiments
  • Applications include HPLC, GC, and MS