Learn More
Diagnostic Biosystems FLUOROMOUNT Fluoromount™
Catalog No. 502407591
Encompass_Preferred
Fluoromount™ is an innovative, aqueous-based mounting medium designed especially for the permanent preservation of fluorescent specimens. Fluoromount™is to be used with tissue sections stained with a multitude of fluorescent dyes. This product is compatible with FITC, phycoerythrin, phycocyanin, and allophycocyanin. It can also be used with other fluorescent markers such as Texas Red and rhodamine.Mounting tissue specimens stained with fluorescent dyes.
Catalog No. 50-240-7591 Supplier Diagnostic Biosystems Supplier No. K024
Fluoromount™ is an innovative, aqueous-based mounting medium designed especially for the permanent preservation of fluorescent specimens. Fluoromount™is to be used with tissue sections stained with a multitude of fluorescent dyes. This product is compatible with FITC, phycoerythrin, phycocyanin, and allophycocyanin. It can also be used with other fluorescent markers such as Texas Red and rhodamine.Mounting tissue specimens stained with fluorescent dyes.