Learn More
Electron Microscopy Sciences Fluoromount-G™ Slide Mounting Medium 25 ML
SDP
Supplier:  Electron Microscopy Sciences 1798425
Encompass_Preferred
A special mounting media for immunofluorescent staining procedures. It is a water soluble, non-fluorescing medium which is well suited for use when the staining procedure has an aqueous final step. It contains 10% polyvinyl alcohol in phosphate buffered glycerol with 0.1% sodium azide as a preservative. One bottle is sufficient to mount up to 500 slides.
Catalog No. 50-259-73