Learn More
Diagnostic Biosystems FLUOROMOUNT/PLUS Fluoromount/Plus™
Catalog No. 502407382
Encompass_Preferred
Fluoromount/Plus™ is an innovative, aqueous-based mounting medium designed especially for the permanent preservation of fluorescent specimens. Fluoromount/Plus™is to be used with tissue sections stained with a multitude of fluorescent dyes. This product is compatible with FITC, phycoerythrin, phycocyanin and allophycocyanin. It can also be used with other fluorescent markers such as Texas Red and rhodamine.\n\nMounting tissue specimens stained with fluorescent dyes.
Catalog No. 50-240-7382 Supplier Diagnostic Biosystems Supplier No. K048200
Fluoromount/Plus™ is an innovative, aqueous-based mounting medium designed especially for the permanent preservation of fluorescent specimens. Fluoromount/Plus™is to be used with tissue sections stained with a multitude of fluorescent dyes. This product is compatible with FITC, phycoerythrin, phycocyanin and allophycocyanin. It can also be used with other fluorescent markers such as Texas Red and rhodamine.\n\nMounting tissue specimens stained with fluorescent dyes.