Learn More
Perkin Elmer US LLC Fluoropolymer Peristaltic Pump Union
Catalog No. 502112217
Encompass_Preferred
Low-flow valve fittings have low dead volume, leak-free integrated ferrule connections.
Catalog No. 50-211-2217 Supplier Perkin Elmer US LLC Supplier No. N0777829
Low-flow valve fittings have low dead volume, leak-free integrated ferrule connections.