Learn More
Aat Bioquest FluoroQuest Anti-fading Mounting Medium with DAPI (20 mL)
SDP
Catalog No. 502249794
Encompass_Preferred
FluoroQuest with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This catalog is for 20 mL.
This item is not returnable. View return policy
Catalog No. 50-224-9794 Supplier Aat Bioquest Supplier No. 20005
FluoroQuest with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This catalog is for 20 mL.