Learn More
Aat Bioquest FluoroQuest Mounting Medium with DAPI (50 mL)
SDP
Catalog No. 502249793
Encompass_Preferred
FluoroQuest with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This catalog is for 50 mL.
This item is not returnable. View return policy
Catalog No. 50-224-9793 Supplier Aat Bioquest Supplier No. 20004
FluoroQuest with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This catalog is for 50 mL.