Learn More
Perkin Elmer US LLC Fluoroscopy Water Standard - 10 mm
Supplier:  Perkin Elmer US LLC L2251293
Encompass_Preferred
Fluoroscopy Water Standards are used to check instrument sensitivity and measure Signal-to-Noise ratios using the Raman band. Compatible with the LS 30/40/50/55 and the FL 6500/8500 spectrometers.
Catalog No. 50-211-2044