Learn More
Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]Octane, Sigma-Aldrich
Catalog No. 01259030 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
20 mL
Catalog No. Volume (Metric)
01-259-030 20 mL
1 options
Catalog No. 01-259-030 Supplier MilliporeSigma Supelco Supplier No. F693720ML
Add to Cart
Add to Cart
Fluoroshield™ with 1,4-Diazabicyclo[2. 2. 2]octane mounting medium is fortified with 1,4-Diazabicyclo[2. 2. 2]octane (a generic version of DABCO™), an anti-fade reagent used to retard photobleaching of fluorescein and other fluorescent dyes.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 20 mL