Learn More
Fluoroshield™ with DAPI, Sigma-Aldrich
Catalog No. 01258870 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
20 mL
Catalog No. Volume (Metric)
01-258-870 20 mL
1 options
Catalog No. 01-258-870 Supplier MilliporeSigma Supelco Supplier No. F605720ML
Add to Cart
Add to Cart
Fluoroshield™ with DAPI has been used:
  • as a counterstain in immunocytochemistry to label nuclei of neural progenitor cells
  • as a histology mounting medium for cardiac tissues sections for immunofluorescence studies
  • as mounting solution for immunostained heart sections for nuclei detection
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 20 mL