Learn More
Fluoroshield™, Sigma-Aldrich
Catalog No. 01258928 Shop All MilliporeSigma Supelco Products
Catalog No. 01-258-928 Supplier MilliporeSigma Supelco Supplier No. F618220ML
Add to Cart
Add to Cart
Fluoroshield is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula not only enables prolonged fluorescence, but also prevents rapid photobleaching (fading) of fluorochromes including FITC, Texas Red™, AMCA, Cy™, Cy3™, Cy5, Alexa Fluor™ 488, Alexa Fluor™ 594, Green fluorescent protein (GFP), tetramethylrhodamine, R-phycoerythrin (R-PE), phycocyanin (PC), and allophycocyanin (APC).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mounting Medium
Reagent Type Mounting Solution