Learn More
Sigma Aldrich Fine Chemicals Biosciences Fluoroshield, 20 mL
Catalog No. NC0375254
Encompass
Used as a mounting medium for cultured megakaryocytes and immunostained platelets for confocal microscopy studies. Used as a mounting medium for 4′,6-diamidino-2-phenylindole (DAPI) stained heart sections for immunofluorescence studies.
Catalog No. NC0375254 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F618220ML
Used as a mounting medium for cultured megakaryocytes and immunostained platelets for confocal microscopy studies. Used as a mounting medium for 4′,6-diamidino-2-phenylindole (DAPI) stained heart sections for immunofluorescence studies.