Learn More
Sigma Aldrich Fine Chemicals Biosciences fluoroshieldTM with DAPI h
Catalog No. 502901819
Encompass
fluoroshield with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This mounting medium is fortified with DAPI which is a counterstain for DNA. for use in in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360 nm and emits at 460 nm producing a blue fluorescence. RNA is also stained with DAPI.
Catalog No. 50-290-1819 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F605720ML
fluoroshield with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This mounting medium is fortified with DAPI which is a counterstain for DNA. for use in in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360 nm and emits at 460 nm producing a blue fluorescence. RNA is also stained with DAPI.