Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorosilicic Acid Standard, 400 ppm, Reagents

Catalog No. 01335193
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-193 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-193 Supplier Reagents Holdings Llc Supplier No. CF1357004A
Add to Cart
Add to Cart

Fluorosilicic Acid Standard, 400 ppm, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Fluorosilicic Acid Standard, 400 ppm
Packaging Poly Bottle
CAS 7732-18-5, 1333-82-0, 7664-93-9
Specific Gravity 1.256
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solubility Completely miscible
Quantity 4 L
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.