Learn More
Mabtech Inc FluoroSpot Plus: Human IFN-γ/IL-2/TNF-α, Pre-coated 96-well PVDF membrane plates, 10 plates
Catalog No. NC2471198
Encompass
The MABTECH FluoroSpot Plus kit is designed for sensitive and multiplexed detection of human biomolecules at a single-cell level, combining the advantages of ELISpot with the capability to analyze multiple analytes simultaneously. Suitable for research use, it supports large-scale screenings and features pre-coated plates for easy assays.
  • Detects human IFN-γ, IL-2, and TNF-α
  • Utilizes FluoroSpot assay method
  • Includes pre-coated 96-well PVDF membrane plates
  • Provides robust assay performance
  • Supports freshly prepared or cryopreserved cells
  • Has a shelf life of at least 12 months
  • Requires storage at 4-8 °C for reagents and room temperature for plates
  • Ships at ambient temperature
  • Offers fluorescent detection capabilities
  • Includes positive control and co-stimulatory antibodies
Catalog No. NC2471198 Supplier Mabtech Inc Supplier No. FSP01020910
Add to Cart
Add to Cart
Request a Quote
The MABTECH FluoroSpot Plus kit is designed for sensitive and multiplexed detection of human biomolecules at a single-cell level, combining the advantages of ELISpot with the capability to analyze multiple analytes simultaneously. Suitable for research use, it supports large-scale screenings and features pre-coated plates for easy assays.
  • Detects human IFN-γ, IL-2, and TNF-α
  • Utilizes FluoroSpot assay method
  • Includes pre-coated 96-well PVDF membrane plates
  • Provides robust assay performance
  • Supports freshly prepared or cryopreserved cells
  • Has a shelf life of at least 12 months
  • Requires storage at 4-8 °C for reagents and room temperature for plates
  • Ships at ambient temperature
  • Offers fluorescent detection capabilities
  • Includes positive control and co-stimulatory antibodies