Learn More
Apexbio Technology LLC FLUOROTHYL-200MG
SDP
Catalog No. 5000568789
Encompass_Preferred
Fluorothyl (CAS No 333-36-8) is a compound predominantly utilized in the field of neurological research originating from the study of fluorinated ethers Its mechanism of action involves the disruption of neural activity through competitive antagonism of GABA A receptors which can initiate seizure activity for the purpose of studying epileptogenesis and anticonvulsant testing In vitro activity is observed in the nanomolar to low micromolar range making it highly potent for research applications Common applications include studies using neuronal cell models and animal studies particularly rodents to evaluate anticonvulsant drugs and understand seizure pathways Experimental concentrations typically depend on the specific research design and desired outcomes Fluorothyl contributes significantly to the understanding of neuronal excitability and the development of therapeutic agents targeting neurological disorders
Catalog No. 50-005-68789 Supplier Apexbio Technology LLC Supplier No. C6616200
Fluorothyl (CAS No 333-36-8) is a compound predominantly utilized in the field of neurological research originating from the study of fluorinated ethers Its mechanism of action involves the disruption of neural activity through competitive antagonism of GABA A receptors which can initiate seizure activity for the purpose of studying epileptogenesis and anticonvulsant testing In vitro activity is observed in the nanomolar to low micromolar range making it highly potent for research applications Common applications include studies using neuronal cell models and animal studies particularly rodents to evaluate anticonvulsant drugs and understand seizure pathways Experimental concentrations typically depend on the specific research design and desired outcomes Fluorothyl contributes significantly to the understanding of neuronal excitability and the development of therapeutic agents targeting neurological disorders