Learn More
Selleck Chemical LLC Fluorouracil (5-Fluorouracil, 5-FU) 100mg 51-21-8 NSC 19893
SDP
Supplier:  Selleck Chemical LLC S1209100MG
Encompass_Preferred
Fluorouracil (5-Fluorouracil, 5-FU, NSC 19893) is a DNA/RNA synthesis inhibitor, which interrupts nucleotide synthetic by inhibiting thymidylate synthase (TS) in tumor cells. Fluorouracil induces apoptosis and can be used in the treatment of HIV. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-816-2