Learn More
- Home
- Products
- Cell Analysis Products
- Microspheres
- Invitrogen™ FluoSpheres™ Carboxylate-Modified Microspheres
Invitrogen™ FluoSpheres™ Carboxylate-Modified Microspheres
Catalog No. F8789
| Catalog No. | Diameter (Metric) | Color | Excitation/Emission | Quantity |
|---|---|---|---|---|
| F8789 | 0.04 μm | Dark Red | 660/680 nm | 1 mL |
| F8803 | 0.1 μm | Yellow-Green | 505/515 nm | 10 mL |
| F8816 | 1.0 μm | Crimson | 625/645 nm | 2 mL |
| F8823 | 1.0 μm | Yellow-Green | 505/515 nm | 10 mL |
| F8801 | 0.1 μm | Red | 580/605 nm | 10 mL |
| F8807 | 0.2 μm | Dark Red | 660/680 nm | 2 mL |
| F10720 | 0.04 μm | Yellow-Green, Orange, Red, Dark Red | 505/515, 540/560, 580/605, 660/680 nm | 1 mL/each |
| F20881 | 0.2 μm | Orange | 365/610 nm | 2 mL |
| F8783 | 0.02 μm | Dark Red | 660/680 nm | 2 mL |
| F8786 | 0.02 μm | Red | 580/605 nm | 10 mL |
| F8795 | 0.04 μm | Yellow-Green | 505/515 nm | 1 mL |
| F8813 | 0.5 μm | Yellow-Green | 505/515 nm | 10 mL |
| F8820 | 1.0 μm | Orange | 540/560 nm | 10 mL |
| F8825 | 2.0 μm | Nile Red | 535/575 nm | 2 mL |
| F8827 | 2.0 μm | Yellow-Green | 505/515 nm | 2 mL |
| F8781 | 0.02 μm | Blue | 365/415 nm | 10 mL |
| F8782 | 0.02 μm | Crimson | 625/645 nm | 2 mL |
| F8784 | 0.02 μm | Nile Red | 535/575 nm | 10 mL |
| F8792 | 0.04 μm | Orange | 540/560 nm | 1 mL |
| F8793 | 0.04 μm | Red | 580/605 nm | 1 mL |
| F8794 | 0.04 μm | Red-Orange | 565/580 nm | 1 mL |
| F8797 | 0.1 μm | Blue | 350/440 nm | 10 mL |
| F8799 | 0.1 μm | Infrared | 715/755 nm | 1 mL |
| F8800 | 0.1 μm | Orange | 540/560 nm | 10 mL |
| F8805 | 0.2 μm | Blue | 365/415 nm | 10 mL |
| F8806 | 0.2 μm | Crimson | 625/645 nm | 2 mL |
| F8809 | 0.2 μm | Orange | 540/560 nm | 10 mL |
| F8810 | 0.2 μm | Red | 580/605 nm | 10 mL |
| F8812 | 0.5 μm | Red | 580/605 nm | 10 mL |
| F8814 | 1.0 μm | Blue | 365/415 nm | 10 mL |
| F8815 | 1.0 μm | Blue | 350/440 nm | 10 mL |
| F8819 | 1.0 μm | Nile Red | 535/575 nm | 10 mL |
| F8821 | 1.0 μm | Red | 580/605 nm | 10 mL |
| F8824 | 2.0 μm | Blue | 365/415 nm | 2 mL |
| F8826 | 2.0 μm | Red | 580/605 nm | 2 mL |
missing translation for 'validMainframeMsg'
Description
Easily perform flow cytometry, microscopy, HTS, HCS, immunoassay, and other laboratory applications using our extensive selection of FluoSpheres Carboxylate-Modified Microspheres. FluoSphere beads can be used in passive adsorption or active, covalent coupling of proteins, nucleic acids, and biomolecules for particle capture applications. FluoSphere microspheres are loaded with proprietary fluorescent dyes, making them the brightest microspheres available.
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C) and protect from light. |
| Color | Dark Red |
| For Use With (Application) | Fluorescence Microscopy |
| Material | Polystyrene |
| Product Type | Carboxylate-Modified Microsphere |
| Excitation/Emission | 660/680 nm |
| Product Line | FLUOSPHERES |
| Quantity | 1 mL |
| Surface Modification | Carboxylate |
| Diameter (Metric) | 0.04 μm |
Frequently Asked Questions (FAQs)
The CML beads have a high density of carboxyl groups at the surface. The surface layer is quite hydrophilic and at the appropriate pH (pH>5), are charged; due to electrostatic repulsion, this type of surface is 3-dimensional and may be considered analogous to the fuzz on a tennis ball.
The smaller the microspheres, the greater the propensity to aggregate. But the aggregation is not irreversible. Sonicate in a bath sonicator or vortex to disperse, just prior to use. You can also add a small concentration of Tween-20 or Triton X-100 (unless you are using them in a live-cell system).
Use of a bath sonicator is recommended to help break up any aggregated microspheres. The foaming is from Tween-20, which is in the stock solution to help prevent aggregation. It is normal and expected to see bubbles from this. Do not use a probe sonicator, which would cause damage to the microspheres (as well as much more bubbling).
The warranty period for FluoSpheres microspheres is 1-year from the date of shipment.
Centrifugation is not an effective way to collect smaller microspheres; many particles remain in the solution even if you can visualize a small pellet. For beads less than 1 µm in diameter, we recommend washing by either:
Cross-flow filtration, as these particles have a very high compression modulus and can withstand high g-forces without risk of harm or dialysis with a 500 kDa MWCO
Note: Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm.
Bacterial contamination is the most common cause of microspheres becoming unusable. Many of our particles are supplied with a low level of sodium azide to prevent bacterial contamination, but sometimes this can still occur. Bacterial contamination is best assessed by plating on appropriate growth medium and checking the plates after 72 hr.
Even brief freezing can cause irreversible aggregation and potential distortion of the bead shape. You should not use these microspheres.
Non-specific binding can often be relieved by a blocking solution, but microspheres seem to require a stronger blocking solution than those most commonly commercially available. Hence, we've developed the BlockAid Blocking Solution (Cat. No. B10710). This reagent is a protein-based blocking solution designed for use with FluoSpheres microspheres and TransFluoSpheres microspheres conjugated to biotin, streptavidin, NeutrAvidin biotin-binding protein, or other proteins. The BlockAid Blocking Solution has proven useful for reducing the nonspecific binding of protein-coated or other macromolecule-coated microspheres in a wide variety of flow cytometry, microscopy, and microarray applications.
We recommend using a bath sonicator to disperse microsphere aggregates. Do not use a probe sonicator as this will damage the microspheres.
The foaming is from Tween 20, which is present in the stock solution to help prevent aggregation. It is normal to see bubbles from this reagent.
Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm, but once pelleted they may not disperse readily or not at all. Centrifugation is not an effective way to collect smaller microspheres; for beads less than 1 µm in diameter, we recommend washing by either cross-flow filtration or dialysis with a 500 kDa MWCO.
We offer a variety of FluoSphere microspheres and kits specifically designed for blood flow determination; if the available colors and/or sizes do not suit your needs, we recommend using beads modified with carboxylate groups for blood flow determination applications. We give some basic usage guidelines in the FluoSpheres Fluorescent Micospheres for Tracer Studies product manual (https://tools.thermofisher.com/content/sfs/manuals/mp13080.pdf). Additionally, the University of Washington Seattle offers the Fluorescent Microsphere Resource Center (FMRC) website (http://fmrc.pulmcc.washington.edu/fmrc.shtml) with detailed applications manuals and references for using fluorescent microspheres to evaluate regional blood flow.
Point spread functions will require a subresolution bead, and we have two different subresolution bead products that could be used for this purpose. TetraSpeck Microspheres, 0.1 µm (Cat. No. T7279), are single subresolution microspheres that are simultaneously stained with blue, green, orange, and dark red dyes. The PS-Speck Microscope Point Source Kit (Cat. No. P7220) consists of individually-stained blue, green, orange, and dark red fluorescent microspheres that are approximately 175 nm in diameter.
We recommend using a ProLong product or any other aqueous mounting media. You should avoid organic-based mountants such as toluene-based mounting media.
The FluoSpheres microspheres may have a trace amount of surfactant. Only IDC Latex beads with diameters greater than 100 nm are actually surfactant-free.
The stability in alcohol or water/alcohol mixtures is dependent on the alcohol chain length. The beads are fairly stable in 40% methanol, but they will dissolve much more readily in long chain alcohols. Prolonged storage of beads in an alcohol containing solution will tend to leach out the dye.
We recommend using dialysis with a MWCO of approximately 500,000 to remove the sodium azide from the microsphere-containing buffer.
We do not determine the exact number of dye molecules per microsphere. The number of dye molecules is a function of the diameter of the particle and the properties of the dye.
The polystyrene microspheres are swelled in a solvent containing a hydrophobic dye specific to the FluoSpheres product. The dye is then able to diffuse into the polystyrene matrix. The beads are then removed from the solvent and dialyzed into an aqueous environment; this reverses the swelling and traps the dye in the polystyrene. Once trapped, the dye is fairly protected from the external environment. The exact protocol and identity of the hydrophobic dyes are proprietary.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.