Learn More
Fluotics Agilent Bravo compatible P250, non-sterile
Catalog No. NC1930545
Encompass
Agilent Bravo compatible P250, non-sterile, working volume: 250 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 19477-002
Catalog No. NC1930545 Supplier Fluotics Supplier No. AGI250.NS
Agilent Bravo compatible P250, non-sterile, working volume: 250 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 19477-002