Learn More
Greiner Bio-One FLUOTRAC™ 200 96-well Non-Treated Microplate
GREENER_CHOICE
Catalog No. 07000085 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
White
Binding Property:
High Binding
Medium Binding
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Binding Property
07-000-085 White Medium Binding
07-000-721 Black Medium Binding
07-000-083 White High Binding
07-000-722 Black High Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-000-085 Supplier Greiner Bio-One Supplier No. 675075
Add to Cart
Add to Cart
For use in fluorescence immunoassay. Greiner Bio-One FLUOTRAC™ 200 96-Well Non-Treated Microplate is suitable for non polar proteins and peptides.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Can be traced back to production through defined LOT number system
  • Reduction of sample volume by up to 50 %
  • Sta
Service Plan
No
Warranty and Services
No
TRUSTED_SUSTAINABILITY
Specifications
Breakapart No
Binding Property Medium Binding
Format Half-Area
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Medium Binding
Color White
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 199 μL
Well Shape Round, Flat Bottom (F-well)
No. per Case 40
No. per Pack 10
Quantity 40/Cs.
RNase Free Yes
Well Color White