Learn More
Greiner Bio-One FLUOTRAC™ 200 HiBase μClear™-Bottom 1536-well Microplate
GREENER_CHOICE
Catalog No. 07000017 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-017 Clear
1 options
Catalog No. 07-000-017 Supplier Greiner Bio-One Supplier No. 782096
Add to Cart
Add to Cart
Free of detectable DNase, RNase, human DNA and pyrogens
Alphanumeric well coding
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens
Format Standard
Lid With Lid
Array 32 x 48
Autoclavable Not Autoclavable
Binding Type Medium Binding
Color Clear
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 3 to 10 μL
Sterility Non-sterile
Well Shape Rounded Square
Dnase Free Yes
Max. RCF 1500 x g (swinging-bucket rotor)
No. per Case 60
No. per Pack 15
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Dimensions (L x W x H) 128 x 85 x 10 mm