Learn More
Greiner Bio-One Fluotrac™ 600 High Binding Flat Bottomed Microplates
GREENER_CHOICE
Catalog No. 07000089 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-089 Black, Clear Bottom
07-001-001 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-089 Supplier Greiner Bio-One Supplier No. 655097
Add to Cart
Add to Cart
Used for throughput screening.
  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Free of detectable DNase, RNase, human DNA
  • Prevents wicking
TRUSTED_SUSTAINABILITY
Specifications
Certified Free of human DNA and pyrogens
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type High Binding
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 0.025 to 0.340 mL
Sterility Sterile
Well Shape Flat Bottom (F-well)
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 10
Quantity 40/Cs.
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Dimensions (L x W x H) 128 x 85 x 14 mm