Learn More
TARGETMOL CHEMICALS INC Fluoxetine 200MG
SDP
Catalog No. 502880685
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Fluoxetine (LY-110140) is a highly specific serotonin uptake inhibitor and selective 5-hydroxytryptamine (5-HT) reuptake inhibitor. Fluoxetine has antidepressant activity. Purity 99.65%
Catalog No. 50-288-0685 Supplier TARGETMOL CHEMICALS INC Supplier No. T0450200MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Fluoxetine (LY-110140) is a highly specific serotonin uptake inhibitor and selective 5-hydroxytryptamine (5-HT) reuptake inhibitor. Fluoxetine has antidepressant activity. Purity 99.65%