Learn More
Medchemexpress LLC Fluoxetine-d5 (hydrochloride) | 1173020-43-3 | 99.9% | C17H14D5ClF3NO | 5 MG
SDP
Catalog No. 5000433357
Encompass_Preferred
Fluoxetine-d5 hydrochloride is a deuterium-labeled form of Fluoxetine hydrochloride. It is a solid, white to off-white compound intended for research use as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules to influence pharmacokinetic and metabolic profiles during drug development. It is suitable for laboratory chemical applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes for drug development quantitation
  • Suitable for laboratory chemical applications
  • Stable under recommended storage conditions
Catalog No. 50-004-33357 Supplier Medchemexpress LLC Supplier No. HYB0102AS5MG
Fluoxetine-d5 hydrochloride is a deuterium-labeled form of Fluoxetine hydrochloride. It is a solid, white to off-white compound intended for research use as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules to influence pharmacokinetic and metabolic profiles during drug development. It is suitable for laboratory chemical applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes for drug development quantitation
  • Suitable for laboratory chemical applications
  • Stable under recommended storage conditions