Learn More
Medchemexpress LLC (±)-Fluoxetine-dd5 oxalate (phenyl-dd5) | 1219804-82-6 | C19H15D5F3NO5 | 1 MG
SDP
Catalog No. 5000373925
Encompass_Preferred
(±)-Fluoxetine-dd5 Oxalate (phenyl-dd5) is the deuterium labeled form of (±)-Fluoxetine Oxalate.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules for quantitation during drug development
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-73925 Supplier Medchemexpress LLC Supplier No. HY151910S11MG
(±)-Fluoxetine-dd5 Oxalate (phenyl-dd5) is the deuterium labeled form of (±)-Fluoxetine Oxalate.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules for quantitation during drug development
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs