Learn More
Enzo Life Sciences Fluoxetine . HCl (50mg). CAS: 56296-78-7
SDP
Catalog No. 502011212
Encompass_Preferred
Potent and selective serotonin uptake inhibitor. Clinically useful antidepressant. Purity: ≥98% (UHPLC). Solubility: Soluble in water (4mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1212 Supplier Enzo Life Sciences Supplier No. BMLNS1400050
Potent and selective serotonin uptake inhibitor. Clinically useful antidepressant. Purity: ≥98% (UHPLC). Solubility: Soluble in water (4mg/ml). Long Term Storage: Ambient.